Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Ferrule for 1/16 in. Tubing
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™ and DISPOSAFLEX™ columns.
Brand:  DWK Life Sciences 420822-0116
Product Code. 11762344
  • 9.50€ / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • EFTE Flangeless Ferrule for 1/16 in. OD Tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferrule
For 0.06 in. O.D. Tubing and Chromaflex Column Chromatography Fitting
ETFE
Case of 1