Learn More
Lonza™ Kinetic-QCL™ Chromogenic LAL Assay Kit
Description
This test utilizes a preparation of Limulus Amebocyte Lysate (LAL), in combination with an 96-well microplate, an absorbance plate reader and appropriate software (such as WinKQCL™) to detect endotoxin photometrically. In the presence of endotoxin, the lysate will begin to cleave the chromogenic substrate, causing the solution to become yellow. The time required for the change is inversely proportional to the amount of endotoxin present.
- The chromogenic nature of the assay makes it the most appropriate choice for testing small volume parenterals, vaccines, antibiotics and biologicals
- It is less impacted by inhibitory products that may interfere with the clotting mechanism in turbidimetric and gel clot assays
- With a sensitivity range of 0.005–50.0 EU/ml, it is the most sensitive of the LAL assays
- Not intended for detection of endotoxin in clinical samples or as an aid in the diagnosis of human disease
Specifications
Specifications
| Includes | Kinetic-QCL™ lysate reagent, E. coli 055:B5 endotoxin and LAL reagent water |
| Quantity | 1 kit |
| Sensitivity | 0.005 to 50.0 EU/mL |
| Total Assay Time | ∼1 hr. |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Gram negative bacterial endotoxin |
| Product Type | Kinetic-QCL Chromogenic LAL Assay Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.