Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand:  HMC Europe 50027
 View more versions of this product
Product Code. 10012832
  • 402.00€ / Each
Explore more special offers
Description
Description
For use with the HMC-HG and HMC-HV autoclaves.
Specifications
Specifications
11 in.
6.6 gal.
HV 25 autoclave
260 mm
Stainless Steel
280 mm
25 L
10.23 in.
With collecting pan
280 x 260 mm