Learn More
HMC Europe Vertical Autoclave Air Cooling RL
Automatic air cooling decreaes turnaround time for busy applications.
Brand: HMC Europe 50002
This item is not returnable.
View return policy
Description
Faster cooling increases workflow in busy labs and is an important safety consideration. Eliminates burnt fingers and dropped and shattered glassware.Specifications
Frequent sterilization | |
HMC-HV autoclave | |
Air Cooling |