Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders with Safety-coated Tube
Brand:  DWK Life Sciences 885460-0022
 View more versions of this product
Product Code. 11813153
  • 325.00€ / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Autoclavable transparent coating
  • With glass pestle
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.16 oz.
0.16 oz., 5mL
8mm
5.9 in.
150mm
22
18mm
0.1 to 0.15mm
0.004 to 0.006 in.
0.31 in.
8.66 in.
220mm
Glass
0.7 in.
5 mL