Learn More
DWK Life Sciences KIMBLE™ EPA Water Analysis Vial, Without Closure
These clear and amber screw thread EPA vials for the collection and storage of water samples comply with the guidelines for establishing test procedures for the analysis of pollutants.
Brand:  DWK Life Sciences 60958A-6
 View more versions of this product
Product Code. 12713077
  • 831.00€ / Pack of 432
Estimated Shipment: 26-01-2026
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear EPA vials are Converted from Type I, 33 expansion borosilicate glass tubing and amber EPA vials are converted from Type I, 51 expansion borosilicate glass tubing
  • While vials are not pre-cleaned, they are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and then are placed into a corrugated shelf-pack
  • Open-top closures and PTFE-faced silicone septa or solid-top PFTE-faced white rubber-lined closures are available separately
Compliance
Vials comply with guidelines for establishing test procedures for analyzing pollutants in water (Ref. U.S. EPA Methods 601, 602, 603, and 604).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.1 in.
28 mm
30 mL
2.75 in.
1.01 oz.
Screw Cap
EPA 601, 602, 603, 604
Clear
Borosilicate Glass
For water analysis
144
30 mL
70 mm
Vial