Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Black phenolic closure with PTFE-lined cap
Brand:  DWK Life Sciences 73802-13425
 View more versions of this product
Product Code. 12184001
  • 174.00€ / Pack of 144
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13-425
Screw Cap
Black
PTFE, Rubber
Phenolic