Learn More
DWK Life Sciences Kimble™ Kontes™ Screw Thread Vials
Combine a low coefficient of expansion with a very high chemical resistance
200.00€
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Combine a low coefficient of expansion with a very high chemical resistance |
Includes | Black phenolic cap and cone-shaped polyethylene |
Thread Style | Screw |
Product Code | Brand | Volume (Metric) | Height (English) | Volume (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Height (English) | Volume (English) | Price | Quantity & Availability | |||
16307528
|
DWK Life Sciences
60941A-8 |
8 mL | 2.48 in. | 0.27 oz. |
200.00€
Pack of 144 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||
Description
- Composed of KIMAX KG-33 borosilicate glass
- Vial top has screw thread finish
- Attached closure has solid top
- Cylindrical shape provides solid balance
- Flat bottom facilitates free-standing capability
Compliance
ASTM Type 1, Class A and USP Type 1 Glass.
Specifications
Clear | |
Combine a low coefficient of expansion with a very high chemical resistance | |
Screw | |
Screw Cap |
Borosilicate Glass | |
Black phenolic cap and cone-shaped polyethylene | |
Phenolic | |
ASTM E438, USP Type I |