missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers Head Only
TLC Sprayer head for use with 125 mL Flask with Knots Sprayer System
Brand: DWK Life Sciences 422531-0125
This item is not returnable.
View return policy
Description
- Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
- Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
- Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
- Adjust spray pattern by varying the position of thumb on vent hole
- Autoclavable (after cap, loosening ring, and O-ring are removed)

Specifications
4.22 oz. | |
Sprayer Head | |
1 Case | |
To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography |
125 mL | |
Glass | |
TLC Reagent Sprayer Head | |
125 mL Flask with Knots Sprayer System |
Product Suggestions
Customers who viewed this item also viewed
KIM001 Kimble Chase 08 09 Catalog
File size: 57.1 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers Head Only > For 125mL flask with Kontes 422530-0125 Sprayer System
Spot an opportunity for improvement?Share a Content Correction