Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Pipets
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
Brand:  DWK Life Sciences 897085-0009
 View more versions of this product
Product Code. 11864065
  • 258.00€ / Pack of 100
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
  • Available with 7 and 9 in. (17.7 and 23cm) tips for use with 7 and 9 in. long NMR tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
100/Cs.
NMR Pipet