Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ Fritted Glass Support Base
The Kimble™ Ultra-Ware™ Fritted Glass Support Base is supplied with a 40/35 joint and glass frit. It is suitable for use with the Kimble™ Ultra-Ware™ 90 mm Microfiltration Assembly.
Brand:  DWK Life Sciences 953826-0090
Product Code. 11893123
  • 1635.00€ / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Product Type: Fritted Support Base
  • Material: Glass
  • Joint Size: 40/35
  • For Use with: Kimble™ Ultra-Ware™ 90 mm Microfiltration Assembly 40/35
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Glass Frit
Glass
40/35
Reusable
Kimble Ultra-Ware 90 mm Microfiltration Assembly 40/35
Non-sterile
Fritted Support Base