Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 608121965
 View more versions of this product
Product Code. 11892983
  • 290.00€ / Pack of 200
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
65 mm
15-425
19 mm
12 mL
3.2 dr.
ASTM Type I, Class A