Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 608121940
 View more versions of this product
Product Code. 11882983
  • 232.00€ / Pack of 200
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
40 mm
15-425
19 mm
6 mL
1.62 dr.
ASTM Type I, Class A