Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 608121545
 View more versions of this product
Product Code. 11852983
  • 215.00€ / Pack of 200
Explore more special offers
Description
Description
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
45 mm
13-425
15 mm
4 mL
1 dr.
ASTM Type I, Class A