Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 608122895
 View more versions of this product
Product Code. 11842993
  • 453.00€ / Pack of 200
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
95 mm
24-400
28 mm
40 mL
10.82 dr.
ASTM Type I, Class A