Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 60812-1235
 View more versions of this product
Product Code. 11832983
  • 207.00€ / Pack of 200
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
35 mm
8-425
12 mm
2 mL
0.5 dr.
ASTM Type I, Class A