Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 60811B-5
 View more versions of this product
Product Code. 11892943
  • 322.00€ / Pack of 72
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Small case quantities
  • Manufactured from clear 33 expansion, borosilicate glass
  • Attached closed top black phenolic closures
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A
Clear
Borosilicate Glass
Screw
Phenolic
57 mm
PTFE/Rubber
28 mm
20 mL
5.4 dr.
Screw Cap (Closed-top)