Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Packaged in corrugated trays with partitions
Brand:  DWK Life Sciences 60811B-3
 View more versions of this product
Product Code. 11852963
  • 402.00€ / Pack of 144
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Small case quantities
  • Manufactured from clear 33 expansion, borosilicate glass
  • Attached closed top black phenolic closures
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A
Clear
Borosilicate Glass
Screw
Phenolic
65 mm
PTFE/Rubber
19 mm
12 mL
3.2 dr.
Screw Cap (Closed-top)