Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached Closures
Attached open top white polypropylene closures
Brand:  DWK Life Sciences 60811S-10
 View more versions of this product
Product Code. 11842953
  • 408.00€ / Pack of 72
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from clear 33 Expansion borosilicate glass
  • Closures supplied with PTFE-faced silicone liners
  • Vials are packaged in corrugated trays with partitions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A
Clear
Borosilicate Glass
Screw
Polypropylene
95 mm
PTFE/Silicone
28 mm
40 mL
10.82 dr.
Screw Cap (Open-top)