Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Sample Vials with Unattached PTFE-Faced/White Rubber Lined Caps
Unattached, closed top black phenolic closures
365.00€ - 480.00€
Specifications
Material (Liner) | PTFE/Rubber |
---|---|
Color | Amber |
Material | Borosilicate Glass |
Thread Style | Screw |
Closure Material | Phenolic |
Product Code | Brand | Closure Type | Volume (English) | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Volume (English) | Volume (Metric) | Price | Quantity & Availability | |||
11873013
|
DWK Life Sciences
60912B2 |
Screw Cap (Closed-top) | 2.16 dr. | 8 mL |
480.00€
Pack of 200 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
11863013
|
DWK Life Sciences
60912B1232 |
Screw Cap | 0.5 dr. | 2 mL |
365.00€
Pack of 200 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
11853013
|
DWK Life Sciences
60912B12 |
Screw Cap | 0.5 dr. | 1.8 mL |
377.00€
Pack of 200 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
11843013
|
DWK Life Sciences
60912B1 |
Screw Cap (Closed-top) | 1 dr. | 4 mL |
404.00€
Pack of 200 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
Description
Unattached, closed top black phenolic closures
- Manufactured from amber 51 expansion borosilicate glass
Specifications
PTFE/Rubber | |
Borosilicate Glass | |
Phenolic |
Amber | |
Screw | |
USP Type I |