Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Manufactured from amber 33 expansion borosilicate glass
Brand:  DWK Life Sciences 609121545
 View more versions of this product
Product Code. 11883003
  • 254.00€ / Pack of 200
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Amber vials offer protection against harmful light and UV rays for light sensitive products.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amber
Borosilicate Glass
Screw
Screw Cap
USP Type I
15 mm
4 mL
1 dr.
45 mm
13-425