Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Product Code. 10257774
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
For Use With (Equipment):
13 mm Plain-end Culture Tubes
16 mm Plain-end Culture Tubes
18 mm Plain-end Culture Tubes
20 mm Plain-end Culture Tubes
25 mm Plain-end Culture Tubes
38 mm Plain-end Culture Tubes
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. 10257774
Brand: DWK Life Sciences 7366418
Only null left
Add to basket
Add to basket
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push Cap
Autoclavable Autoclavable
Material Polypropylene
Color Yellow
Sterility Non-sterile
For Use With (Equipment) 18 mm Plain-end Culture Tubes
Disposable Reusable