missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KHS 101 hydrochloride
GREENER_CHOICE

Product Code. p-5545846
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18712709

Brand: Tocris Bioscience™ 4888/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inducer of neuronal differentiation in hippocampal neural progenitors

KHS 101 hydrochloride is a selective inducer of neuronal differentiation; induces neuronal differentiation in cultured hippocampal neural progenitor cells (NPCs) by interacting with TACC3 (EC50 ∽1 μM). Suppresses astrocyte formation. Also induces acceleration of neuronal differentiation in the hippocampal dentate gyrus in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type KHS 101 hydrochloride
Purity >98%
Molecular Weight (g/mol) 375.92
Inhibitors KHS 101 hydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.