Learn More
Novus Biologicals™ Keratinocyte Differentiation Factor 1 Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-05395
Product Code. 18056866
  • 700.00€ / 0.10mg
Explore more special offers
Description
Description
The Keratinocyte Differentiation Factor 1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_689578. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens C1orf172, mRNA.
Native