missing translation for 'onlineSavingsMsg'
Learn More

kb NB 142-70, Tocris Bioscience™

Product Code. 18074672
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective PKD inhibitor; analog of CID 755673 (Cat. No. 3327)

Selective protein kinase D (PKD) inhibitor (IC50 values are 28.3, 58.7 and 53.2 nM for PKD1, 2 and 3 respectively). More potent analog of CID 755673 (Cat. No. 3327) with 7-fold greater inhibition. Inhibits prostrate cancer cell migration and invasion and reduces wound healing in vitro; displays prominent cytotoxic and anti-proliferative effects.

Chemical Identifiers

CAS 1233533-04-4
Molecular Formula C11H9NO2S2
Molecular Weight (g/mol) 251.318
InChI Key DHUAGGSHTKPOHU-UHFFFAOYSA-N
Synonym 3,4-dihydro-9-hydroxy-1 benzothieno 2,3-f-1,4-thiazepin-5 2h-one, 9-hydroxy-3,4-dihydro-2h-1 benzothiolo 2,3-f 1,4 thiazepin-5-one, 3,4-dihydro-9-hydroxy-1 benzothieno-2,3-f-1,4-thiazepin-5 2h-one, 4-hydroxy-8,14-dithia-11-azatricyclo 7.5.0.0 2 ,? tetradeca-1 9 ,2,4,6-tetraen-10-one, qcr-15, kb-nb hplc, 9-hydroxy-3,4-dihydro-2h-1-benzothiolo 2,3-f 1,4 thiazepin-5-one, 9-hydroxy-3,4-dihydro 1 benzothieno 2,3-f 1,4 thiazepin-5 2h-one
PubChem CID 45258277
IUPAC Name 9-hydroxy-3,4-dihydro-2H-[1]benzothiolo[2,3-f][1,4]thiazepin-5-one
SMILES C1CSC2=C(C(=O)N1)SC3=C2C=C(C=C3)O

Specifications

Quantity 50 mg
Formula Weight 251.32
Percent Purity >98%
Chemical Name or Material kb NB 142-70
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'