Learn More
SI Analytics™ Karl Fischer Titration Vessel
SI Analytics™ Karl Fischer Titration Vessel is constructed of DURAN™ glass and comes equipted with screw threads, a titration tip, and more.
Brand:  SI Analytics™ 285216677
 View more versions of this product
Product Code. 10598221
  • 1350.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes a removable head made of polypropylene/PTFE
  • Includes 1 drilling NS 19, NS 14.5, NS 7.5 and 3 drillings with screw threads
  • Includes a titration tip, moisture trap and weighing funnel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Titration Vessel
Removable head, moisture trap, and weighing funnel
DURAN™ Glass
30 to 150 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ Karl Fischer Titration Vessel > Capacity: 30 to 150mL