Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand:  Poeppelmann™ 61000160000
 View more versions of this product
Product Code. 10451405
  • 22.05€ / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances
Specifications
Specifications
Yellow
Used universally as plug or cap, Tapered design is ideal for accommodating tolerances
100 Pack
Polyethylene
17.5 mm
15 mm
24 mm
Closing Plug