missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ Kapsto™ Polyethylene Tapered Closures

Manufactured from polyethylene. Poppelmann™ Kapsto™ Polyethylene Tapered Closures can be used universally as plug or cap.

143.00€

Specifications

Product Type Tapered Closure
Material Polyethylene
For Use With (Application) Can be used universally as plug or cap
Packaging Quantity 100 Pack
Color Natural
View More Specs

Products 1
Product Code Brand Overall Length (Metric) Price Quantity & Availability  
Product Code Brand Overall Length (Metric) Price Quantity & Availability  
10410075
View Documents
Poeppelmann™
60006980000
13 mm
143.00€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Polyethylene
  • Yellow
Specifications

Specifications

Tapered Closure
Can be used universally as plug or cap
Natural
Polyethylene
100 Pack
Videos
SDS
Documents

Documents

Product Certifications