missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs

Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.

12.35€ - 35.10€

Specifications

Material Polyethylene
For Use With (Application) Can be used universally as plug or cap. The tapered design is ideal for accommodating tolerances
Packaging Quantity 100 Pack
Color Yellow
Product Type Sealing Plug
View More Specs

Products 2
Product Code Brand Overall Length (Metric) Diameter (Metric) Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Overall Length (Metric) Diameter (Metric) Diameter (Metric) Inner Price Quantity & Availability  
10043764
View Documents
Poeppelmann™
61000180000
15 mm 25 mm 17.4 mm
12.35€
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
10595854
View Documents
Poeppelmann™
61000240000
19 mm 33 mm 23.7 mm
35.10€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances
Specifications

Specifications

Polyethylene
100 Pack
Sealing Plug
Can be used universally as plug or cap. The tapered design is ideal for accommodating tolerances
Yellow
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.