missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 1 Antibody (105), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89474PE
This item is not returnable.
View return policy
Description
Kallikrein 1 Monoclonal antibody specifically detects Kallikrein 1 in Human samples. It is validated for ELISA
Specifications
| Kallikrein 1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 105 | |
| ELISA | |
| EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, Klk6, KLKR, Tissue kallikrein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 1 (Accession#: NP_002248.1; Met 1-Ser 262). | |
| 0.1 mL | |
| Cancer | |
| 3816 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction