Learn More
BUCHI K-439 Model Speed Digestor
Brand:  BUCHI 1154392100
Product Code. 11987228
  • 9955.00€ / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Specifications
Speed Digestor
220/240 V ±10 V
620 mm
Feed - Kjeldahl, Chemicals/Agricultural, Beverage, Pharmaceutical / Cosmetics , Food , Environmental Analysis
310 mm
540 mm
50/60 Hz
2000 W
220/240 V ±10 V, 50/60 Hz, 1280 W
300mL Sample tubes and standard suction module
1 Ea.