Learn More
BUCHI K-439 Model SpeedDigestor
The SpeedDigester K-439 revolutionizes IR digestion by combining the benefits of IR and block digestion in one unit, with fast nitrogen determination by Kjeldahl digestion at controlled temperatures enabling an increased sample throughput.
Brand:  BUCHI 1154392200
Product Code. 11997228
  • 10480.00€ / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Fast heat transfer of infrared heaters to samples
  • Large time savings due to fast heating and cooling
  • Increased sample throughput due to short process time
  • Accelerated digestion step with continuous addition of H&sub2;O&sub2;
  • One instrument for both Kjeldahl and reflux digestion
  • All BUCHI samples tubes can be used (100mL, 300mL, 500mL)
  • Sample tubes compliant with ISO 6060 for COD and other reflux digestion
  • Optional dedicated suction mode for aqueous samples
  • Accurate temperature control following the programmed profile
  • Storage of up to 50 different method profiles (time/temperature)
  • Digestion process shown graphically
  • Thermal homogeneity due to the innovative insulation plate
Specifications
Specifications
Speed Digestor
220/240 V ±10 V
620 mm
Feed - Kjeldahl, Chemicals/Agricultural, Beverage, Pharmaceutical / Cosmetics , Food , Environmental Analysis
310 mm
540 mm
50/60 Hz
2000 W
220/240 V ±10 V, 50/60 Hz, 1280 W
100mL, 300mL, 500mL Sample tubes and suction module with condensate trap
1 Ea.