Learn More
BUCHI K-415 Model DuoScrub Scrubber
Protects the user and the environment not only by neutralizing poisonous fumes and reaction gases but also by minimizing the use of cooling water.
Brand:  BUCHI 114152320
Product Code. 13425539
  • 6615.00€ / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • No exposure of the operator to harmful acid fumes
  • Maximum safety in the workplace from a self-contained design
  • Adjustable suction power to control sample dryness
  • Most efficient neutralization due to the large gas/liquid contact area
  • Automatic cooling water control provided for all Scrubber models
  • Full control of the Scrubber via the digester
  • No corrosion of the fume hood
  • Maximal neutralization efficiency by the swirl discs
  • Easy evaporation of water samples (TKN)
  • Broad range of applications due to the four cleaning steps: pre-condensation of fumes, neutralization of acid or alkaline fumes, adsorption of organic or inorganic substances, and redox-reactions of specific fumes
Specifications
Specifications
DuoScrub Scrubber
230 V
526 x 248 x 412mm
412 mm
248 mm
50/60 Hz
526 mm
230V, 50/60 Hz
1 Ea.