missing translation for 'onlineSavingsMsg'
Learn More

K 114, Tocris Bioscience™

Product Code. 18054242
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Amyloid fibril-specific fluorescent dye

Potent amyloid fibril-specific fluorescent dye (EC50 = 20 - 30 nM). Exhibits minimal fluorescence in aqueous buffers and fluoresces brightly in the presence of Aβ, α-synuclein and tau in situ. (Optimum wavelength = 550 nm).

Chemical Identifiers

CAS 872201-12-2
Molecular Formula C22H17BrO2
Molecular Weight (g/mol) 393.28
InChI Key OXPHQQMZTXMEGO-RJTULKDBSA-N
Synonym e,e-1-bromo-2,5-bis-4-hydroxystyryl benzene, k hplc , powder, trans,trans-1-bromo-2,5-bis-4-hydroxystyryl benzene, trans,trans-1-bromo-2,5-bis-4-hydroxy styrylbenzene, 4,4'-2-bromo-1,4-phenylene di e ethene-2,1-diyl diphenol, 4,4'-2-bromo-1,4-phenylene di-1e-2,1-ethenediyl bisphenol, 4-e-2-3-bromo-4-e-2-4-hydroxyphenyl ethenyl phenyl ethenyl phenol
PubChem CID 44828481
ChEBI CHEBI:78674
IUPAC Name 4-[(E)-2-[3-bromo-4-[(E)-2-(4-hydroxyphenyl)ethenyl]phenyl]ethenyl]phenol
SMILES C1=CC(=CC=C1C=CC2=CC(=C(C=C2)C=CC3=CC=C(C=C3)O)Br)O

Specifications

Quantity 50 mg
Formula Weight 393.27
Percent Purity >98%
Chemical Name or Material K 114
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'