missing translation for 'onlineSavingsMsg'
Learn More
Learn More
K 114, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent amyloid fibril-specific fluorescent dye (EC50 = 20 - 30 nM). Exhibits minimal fluorescence in aqueous buffers and fluoresces brightly in the presence of Aβ, α-synuclein and tau in situ. (Optimum wavelength = 550 nm).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 393.27 |
Percent Purity | >98% |
Chemical Name or Material | K 114 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'