Learn More
Justrite™ Safety Fill Vent
Brand:  Justrite™ 08204
 View more versions of this product
Product Code. 15173607
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Specifications
FM
Facilitate Waste Solvent Disposal
6 in. Flame arrester, automatic pressure and vacuum relief and cap with chain
Fill Drum Vent
Brass