Learn More
ITS Thorsten Betzel™ WaterWorks™ 4-in-1 City Water Check Strip
Brand:  ITS Europe 480113
Product Code. 11702967
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Order Info
ForUseWithEquipment
Specifications
Specifications
Strip
25 Pack
WaterWorks 4-in-1 City Water Check Strip
Test for pH, Total Alkalinity, Total Chlorine, and Total Hardness
35 sec.