missing translation for 'onlineSavingsMsg'
Learn More

iso-PPADS tetrasodium salt, Tocris Bioscience™

Product Code. 30001112
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective P2X antagonist

P2X-purinoceptor antagonist (pKi = 6.5) with similar activity to PPADS .

2'4'-isomer also available.

Chemical Identifiers

CAS 207572-67-6
Molecular Formula C14H14N3Na4O12PS2
Molecular Weight (g/mol) 603.328
InChI Key XPRWSDRXOGVWIR-WCZAJHAHSA-N
Synonym Pyridoxalphosphate-6-azophenyl-2', 5'-disulfonic acid tetrasodium salt
PubChem CID 131851157
IUPAC Name 2-[(2Z)-2-[4-formyl-6-methyl-5-oxo-3-(phosphonooxymethyl)pyridin-2-ylidene]hydrazinyl]benzene-1,4-disulfonic acid;sodium
SMILES CC1=NC(=NNC2=C(C=CC(=C2)S(=O)(=O)O)S(=O)(=O)O)C(=C(C1=O)C=O)COP(=O)(O)O.[Na].[Na].[Na].[Na]

Specifications

Quantity 10 mg
Formula Weight 599.3
Percent Purity >98%
Chemical Name or Material iso-PPADS tetrasodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'