missing translation for 'onlineSavingsMsg'
Learn More

ISO 1, Tocris Bioscience™

Product Code. 30002044
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Macrophage migration inhibitory factor (MIF) inhibitor

ISO 1 is a macrophage migration inhibitory factor (MIF) inhibitor (IC50 = 7 μM); inhibits MIF tautomerase activity in vitro and in vivo. Protective against mouse models of streptozotocin -induced diabetes mellitus; blocks anti-inflammatory response following LPS exposure and increases survival. Inhibits airway remodeling in a mouse model of chronic asthma. Cell permeable; orally bioavailable.

Chemical Identifiers

CAS 478336-92-4
Molecular Formula C12H13NO4
Molecular Weight (g/mol) 235.239
InChI Key CLVQGFPQFNASJH-UHFFFAOYSA-N
Synonym iso-1, mif antagonist, iso-1, methyl 2-3-4-hydroxyphenyl-4,5-dihydroisoxazol-5-yl acetate, 5-isoxazoleacetic acid, 4,5-dihydro-3-4-hydroxyphenyl-, methyl ester, macrophage migration inhibitory factor antagonist, iso-1, s,r-3-4-hydroxyphenyl-4,5-dihydro-5-isoxazole acetic acid, methyl ester, methyl 2-3-4-oxocyclohexa-2,5-dien-1-ylidene-1,2-oxazolidin-5-yl acetate, 3-4-hydroxyphenyl-2-isoxazoline-5-acetic acid methyl ester, methyl2-3-4-hydroxyphenyl-4,5-dihydroisoxazol-5-yl acetate, 3-4-hydroxyphenyl-4,5-dihydro-5-isoxazoleacetic methyl ester
PubChem CID 6098948
IUPAC Name methyl 2-[3-(4-oxocyclohexa-2,5-dien-1-ylidene)-1,2-oxazolidin-5-yl]acetate
SMILES COC(=O)CC1CC(=C2C=CC(=O)C=C2)NO1

Specifications

Color White
Quantity 50 mg
Formula Weight 235.24
Percent Purity >98%
Physical Form Solid
Chemical Name or Material ISO 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.