Learn More
Bollé Safety IRI-s Platinum™ Clear Safety Glasses
Manufactured by Bolle Safety the IRI-s Safety Glasses are coated with Platinum™, which is a permanent coating that gives a high scratch resistance and delays fogging beyond the minimum required standards.
Brand: Bollé Safety IRIPSI
108.49 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- PLATINUM™ anti-fog and anti-scratch coating
Specifications
Anti-fog, Scratch-resistant | |
Clear | |
Safety glasses | |
27 g |