missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium ICP & ICP-MS Standard, 10 mg/L Ir in HCl 2%, Honeywell Fluka™
Brand: Honeywell Fluka E1394-100ML
This item is not returnable.
View return policy
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Iridium ICP and ICP-MS Standard | |
Standards for ICP and ICP-MS | |
<2 | |
100 mL |