Learn More
Iridium AAS Standard, 1000 mg/L Ir in HCl10%, Honeywell Fluka™
Brand:  Honeywell Fluka E4036-500ML
 View more versions of this product
Product Code. 16654324
  • 721.78€ / 500mL
Explore more special offers
Description
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Iridium AAS Standard
1.02 g/cm3
<2
500 mL