Learn More
SI Analytics™ IonLine pH Combination Electrode
SI Analytics™ IonLine pH Combination Electrode is a micro-electrode with ID function, temperature sensor, and a fixed cable.
Brand:  SI Analytics™ 285114300
Product Code. 10523524
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Temp. sensor Pt 1000
  • 1 m cable with DIN and banana plug
  • Glass shaft
  • Pt diaphragm
  • KCl 3 mol/l
  • Iodine/Iodide-ref. system
  • Cylindrical membrane
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A-Glass
-5°C to +100°C
40 to 130 mm
12 mm
0 to 14
Micro-Electrode
1 m
5 mm sensor