Learn More
SI Analytics™ Ion-selective Indicator Electrode
With Ag/AgCl-Reference System
Brand:  SI Analytics™ 285216314
Product Code. 10377072
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Calcium electrode
  • Fixed cable
  • DIN plug
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.02 to 40,000 mg/L
Calcium electrode
1 m fixed cable with DIN plug
2.5 to 11
0°C to 40°C