missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Solution, 0.05M, Chem-Lab
Click to view available options
Quantity:
500 mL
1 L
Description
Iodine is a nonmetallic element of the halogen group. It is a nutritionally essential element, especially important in thyroid hormone synthesis. In solution, it has anti-infective properties (when used topically).
Specifications
pH | 5.8–6.2 |
Quantity | 1 L |
IUPAC Name | molecular iodine |
Formula Weight | 253.81g/mol |
Concentration or Composition (by Analyte or Components) | 0.05mol/L |
Density | 1.02g/mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'