missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Iodine Determination Flasks with Conical Ground Joint, without Collar

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

256.00€ - 447.00€

Specifications

Material Duran Glass
No. per Pack 10/Pk.
Standard Taper Joint NS 29/32
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10040311
View Documents
Lenz Laborglasintrumente
03026070
1 L
447.00€
Pack of 10
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
 
10718651
View Documents
Lenz Laborglasintrumente
03026052
300 mL
256.00€
Pack of 10
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
 
Description

Description

  • With hollow hexagonal stopper NS 29/32
  • With scale (approx. volume /ml)
  • With inscription label
  • Material: DURAN™
Specifications

Specifications

Duran Glass
10/Pk.
NS 29/32
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.