Learn More
Invitrogen™ INVSc1, S. cerevisiae Yeast Strain
Fast-growing diploid strain excellent for expression
Brand: Invitrogen™ C81000
This item is not returnable.
View return policy
Description
- INVSc1 has genotype INVSc1: MATa his3D1 leu2 trp1-289 ura3-52 MAT his3D1 leu2 trp1-289 ura3-52
Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Yeast Expression
Specifications
Yeast Strain | |
Yeast | |
1 Stab |
INVSc1 | |
S. cerevisiae |
For Research Use Only. Not for use in diagnostic procedures.