missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pLenti6/V5-DEST™ Gateway™ Vector

Product Code. 11590836
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
This item is not returnable. View return policy

Product Code. 11590836

Brand: Invitrogen™ V49610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

pLenti6/V5-DEST: 6μg (40μL of 150ng/μL vector in 10mM Tris-HCl, 1mM EDTA, pH 8.0); pLenti6/V5-GW/lacZ control: 10μg (20μL of 0.5μg/μL vector in 10mM Tris-HCl, 1mM EDTA, pH 8.0); Stbl3 Chemically Competent E. coli

For lentiviral-based expression of a target gene in dividing and nondividing mammalian cells

  • The vector has CMV promoter for driving constitutive expression of the target gene and the blasticidin selection marker for stable selection
  • C terminal V5 tag for quick detection

Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Promoter CMV
Product Type Lentiviral Expression Vector
Selection Agent (Eukaryotic) Blasticidin
Content And Storage ViraPower™ Lentiviral Gateway™ Vectors, store at –20°C includes:

pLenti6⁄V5-DEST™: 6 μg (40 μl of 150 ng⁄μl vector in 10 mM
Tris-HCl, 1mM EDTA, pH 8.0)

pLenti6⁄V5-GW⁄lacZ control: 10 μg
(20 μl of 0.5 μg⁄μl vector in 10 mM
Tris-HCl, 1mM EDTA, pH 8.0)

One Shot™ Stbl3™ Chemically Competent E. coli store at –80°C
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 6 μg
Vector pDEST, pLenti
Product Line Gateway, ViraPower
For Use With (Application) Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.