missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ pDEST™14 Vector
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
Descripción
Includes
All destination vectors are provided lyophilized and supercoiled.
Requires
Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
- Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Bacterial Expression,Cloning,Gateway Cloning,Protein Expression,Proteins, Expression, Isolation and Analysis
Especificaciones
Especificaciones
| Constitutive or Inducible System | Inducible |
| Promoter | T7 |
| Product Type | Expression Vector |
| Selection Agent (Eukaryotic) | None |
| Content And Storage | All destination vectors are provided lyophilized and supercoiled. |
| Antibiotic Resistance Bacterial | Ampicillin (AmpR) |
| Protein Tag | Untagged |
| Cloning Method | Gateway |
| Quantity | 6 μg |
| Vector | pDEST, Gateway T7 Vectors |
| Mostrar más |
For Research Use Only. Not for use in diagnostic procedures.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido