missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin / IRDN Mouse, Clone: IRDN/794, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3630-MSM4-P1
This item is not returnable.
View return policy
Specifications
| Insulin / IRDN | |
| Cytoplasmic | |
| IRDN/794 | |
| Unconjugated | |
| NP_001172027 | |
| Mouse | |
| 6kDa | |
| Protein A/G Purified | |
| RUO | |
| 3630 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| INS | |
| Recombinant INS protein | |
| MIA PaCa-2 cells or Pancreas | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction